missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Nitroquinoline N-oxide

Numéro de catalogue. N81415G
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
N81415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. N81415G Fournisseur Sigma Aldrich Code fournisseur N81415G
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Nitroquinoline N-oxide

Spécifications

CAS 56-57-5
Melting Point 154°C to 156°C (lit.)
Molecular Formula C9H6N2O3
Linear Formula C9H6N2O3
MDL Number MFCD00006738
Quantity 5 g
Synonym 4-NQO; 4-Nitroquinoline 1-oxide
Molecular Weight (g/mol) 190.16
Percent Purity ≥98%