missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalonitrile, 99%

Catalog No. AC416150250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416150250 25 g Glass bottle
AC416150050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416150250 Supplier Thermo Scientific Chemicals Supplier No. 416150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31643-49-9 | C8H3N3O2 | 173.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31643-49-9
Molecular Formula C8H3N3O2
Molecular Weight (g/mol) 173.13
MDL Number MFCD00040301
InChI Key NTZMSBAAHBICLE-UHFFFAOYSA-N
Synonym 4-nitrophthalonitrile, 5-nitrobenzene-1,2-dicarbonitrile, 4-nitro-1,2-benzenedicarbonitrile, 1,2-dicyano-4-nitrobenzene, 3,4-dicyanonitrobenzene, phthalonitrile, 4-nitro, 4-nitro-phthalonitrile, 1,2-benzenedicarbonitrile, 4-nitro, 5-nitro-1,2-benzenedicarbonitrile, 4-nitro-o-benzenedinitrile
PubChem CID 97443
IUPAC Name 4-nitrobenzene-1,2-dicarbonitrile
SMILES [O-][N+](=O)C1=CC=C(C#N)C(=C1)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 140°C to 144°C
Color Beige to Gray or Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 173.13
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Nitrophthalonitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 250-748-9
RTECSNumber TI8576000
TSCA TSCA

RUO – Research Use Only