missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophthalic Acid 98.0+%, TCI America™

Catalog No. N0244500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0244500G 500 g
N024425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0244500G Supplier TCI America Supplier No. N0244500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610-27-5
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007252
InChI Key SLBQXWXKPNIVSQ-UHFFFAOYSA-N
Synonym 4-nitrophthalic acid, 5-nitrophthalic acid, 4-nitrobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, 4-nitro, phthalic acid, 4-nitro, 4-nitrophthalicacid, p-nitrophthalic acid, 4-nitro-phthalic acid, acmc-209mo8
PubChem CID 69121
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 500 g
Formula Weight 211.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophthalic Acid
EINECSNumber (3)-3666
TSCA Yes