missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenylboronic acid, 97%

Catalog No. AC433890010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433890010 1 g Glass bottle
AC433890050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC433890010 Supplier Thermo Scientific Chemicals Supplier No. 433890010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24067-17-2 | C6H6BNO4 | 166.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24067-17-2
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161360
InChI Key NSFJAFZHYOAMHL-UHFFFAOYSA-N
Synonym 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
PubChem CID 2773552
IUPAC Name (4-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 270°C to 271°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.93
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only