missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Trifluoromethanesulfonate 99.0+%, TCI America™

Catalog No. N09935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N09935G Supplier TCI America Supplier No. N09935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 17763-80-3
Melting Point 54°C
Color Yellow
Molecular Formula C7H4F3NO5S
Quantity 5 g
MDL Number MFCD00192354
UN Number 3261
Synonym trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
InChI Key NDTIXHNCNLKURN-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1
IUPAC Name 4-nitrophenyl trifluoromethanesulfonate
Molecular Weight (g/mol) 271.17
PubChem CID 549870
Formula Weight 271.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Trifluoromethanesulfonate
Show More Show Less