missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific™

Catalog No. p-3770835
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425790010 1g Glass bottle
AC425790050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425790010 Supplier Thermo Scientific Chemicals Supplier No. 425790010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17763-80-3
Molecular Formula C7H4F3NO5S
Molecular Weight (g/mol) 271.17
MDL Number MFCD00192354
InChI Key NDTIXHNCNLKURN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
PubChem CID 549870
IUPAC Name (4-nitrophenyl) trifluoromethanesulfonate
SMILES [O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1g
Formula Weight 271.17
Percent Purity 99%
Chemical Name or Material 4-Nitrophenyl trifluoromethanesulfonate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only