missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl Isothiocyanate 99.0+%, TCI America™

Catalog No. N08265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08265G 5 g
N082625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08265G Supplier TCI America Supplier No. N08265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2131-61-5
Molecular Formula C7H4N2O2S
Molecular Weight (g/mol) 180.181
MDL Number MFCD00007307
InChI Key NXHSSIGRWJENBH-UHFFFAOYSA-N
Synonym 4-nitrophenyl isothiocyanate, 4-nitrophenylisothiocyanate, p-nitrophenyl isothiocyanate, benzene, 1-isothiocyanato-4-nitro, isothiocyanic acid 4-nitrophenyl ester, 1-isothiocyanato-4-nitro-benzene, 4-nitrobenzenisothiocyanate, 4-nitrophenyl isothiocyanate 25gr, pubchem13428, 4nitrophenyl isothiocyanate
PubChem CID 75044
IUPAC Name 1-isothiocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=S)[N+](=O)[O-]

Specifications

Melting Point 112°C
Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 2811
Formula Weight 180.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl Isothiocyanate