missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl alpha-L-Fucopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 10 mg |
| Formula Weight | 285.25 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrophenyl alpha-L-Fucopyranoside |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |