missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl alpha-D-Galactopyranoside 98.0+%, TCI America™

Catalog No. N0492200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0492200MG 200 mg
N04921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0492200MG Supplier TCI America Supplier No. N0492200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7493-95-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00065050
InChI Key IFBHRQDFSNCLOZ-IIRVCBMXSA-N
Synonym 4-nitrophenyl a-d-galactopyranoside, 4-nitrophenyl-alpha-d-galactopyranoside, pnpalphagal, p-nitrophenyl-alpha-d-galactopyranoside, 4-nitrophenyl, a-d-galactopyranoside, p-nitrophenyl alpha-d-galactopyranoside, 4-nitrophenyl alpha-d-galactopyranoside, p-nitrophenyl-alpha-d-galactoside, 4-nitrophenyl-alpha-d-galactoside, 1-o-4-nitrophenyl-alpha-d-galactose
PubChem CID 82000
ChEBI CHEBI:546840
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 200 mg
Formula Weight 301.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl alpha-D-Galactopyranoside [Substrate for alpha-D-Galactosidase]
TSCA No
Recommended Storage Refrigerator