missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-alpha-D-galactopyranoside, 98+%

Catalog No. AAL13376ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13376ME 500 mg
AAL13376MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13376ME Supplier Thermo Scientific Chemicals Supplier No. L13376ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7493-95-0 | C12H15NO8 | 301.251 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7493-95-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00065050
InChI Key IFBHRQDFSNCLOZ-IIRVCBMXSA-N
Synonym 4-nitrophenyl a-d-galactopyranoside, 4-nitrophenyl-alpha-d-galactopyranoside, pnpalphagal, p-nitrophenyl-alpha-d-galactopyranoside, 4-nitrophenyl, a-d-galactopyranoside, p-nitrophenyl alpha-d-galactopyranoside, 4-nitrophenyl alpha-d-galactopyranoside, p-nitrophenyl-alpha-d-galactoside, 4-nitrophenyl-alpha-d-galactoside, 1-o-4-nitrophenyl-alpha-d-galactose
PubChem CID 82000
ChEBI CHEBI:546840
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Melting Point 166°C to 169°C
Quantity 500 mg
Beilstein 92214
Optical Rotation +242° (c=1 in Water)
Formula Weight 301.26
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-alpha-D-galactopyranoside
EINECSNumber 231-343-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only