missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl acetate, 97%

Catalog No. AAL0031409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0031409 10 g
AAL0031418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0031409 Supplier Thermo Scientific Chemicals Supplier No. L0031409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol

4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl acetate is used with iodoacetic acid for reductive cleavage of methionine-containing peptides. It is also used as a substrate that has been used in assays for esterase and lipase activity. Inorganic complexes have been evaluated for their methanolysis or hydrolysis activity using p-nitrophenyl acetate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 830-03-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007326
InChI Key QAUUDNIGJSLPSX-UHFFFAOYSA-N
Synonym p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl ester
PubChem CID 13243
ChEBI CHEBI:82635
IUPAC Name (4-nitrophenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 77°C to 79°C
Quantity 10 g
Beilstein 515874
Solubility Information Insoluble in water.
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl acetate
Hazard Category H317-H318
Precautionary Statement P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
EINECSNumber 212-593-5
RTECSNumber AJ1150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only