missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. N0866200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0866200MG 200 mg
N08661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0866200MG Supplier TCI America Supplier No. N0866200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3459-18-5
Molecular Formula C14H18N2O8
Molecular Weight (g/mol) 342.30
MDL Number MFCD00063696
InChI Key OMRLTNCLYHKQCK-JQXIRTQVNA-N
Synonym 4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamide
PubChem CID 102416
IUPAC Name N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 205°C
Color White-Yellow
Quantity 200 mg
Formula Weight 342.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
TSCA No
Recommended Storage Freezer