missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenethyl alcohol, 99%

Catalog No. AC179610500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC179610500 50 g
AC179610100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC179610500 Supplier Thermo Scientific Chemicals Supplier No. 179610500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-27-6 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

Melting Point 56.0°C to 57.0°C
Boiling Point 148.0°C to 149.0°C (2.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Linear Formula O2NC6H4CH2CH2OH
Quantity 50 g
Beilstein 06,II,238
Formula Weight 167.16
Percent Purity 99%
Chemical Name or Material 4-Nitrophenethyl alcohol
EINECSNumber 202-835-8

RUO – Research Use Only