missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroguaiacol 98.0+%, TCI America™

Catalog No. N08925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08925G 5 g
N089225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08925G Supplier TCI America Supplier No. N08925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3251-56-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.136
MDL Number MFCD00012143
InChI Key IZLVFLOBTPURLP-UHFFFAOYSA-N
Synonym 4-nitroguaiacol, phenol, 2-methoxy-4-nitro, guaiacol, 4-nitro, 2-hydroxy-5-nitroanisole, 3-nitro-6-hydroxyanisole, 4-hydroxy-3-methoxynitrobenzene, phenol, o-methoxy-p-nitro, o-methoxy-p-nitrophenol, 2-methoxy-4-nitro-phenol, 4-nitrogualacol
PubChem CID 76738
ChEBI CHEBI:81050
IUPAC Name 2-methoxy-4-nitrophenol
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])O

Specifications

Melting Point 103°C
Quantity 5 g
Formula Weight 169.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitroguaiacol
RTECSNumber SL7800000
TSCA Yes