missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloride, 99%

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol

Supplier:  Thermo Scientific Chemicals A1574918

 View more versions of this product

Catalog No. AAA1574918

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitrobenzyl chloride
70°C to 74°C
MFCD00007374
UN3261
4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
C1=CC(=CC=C1CCl)[N+](=O)[O-]
171.58
CHEBI:87406
99%
100-14-1
C7H6ClNO2
50 g
387187
KGCNHWXDPDPSBV-UHFFFAOYSA-N
1-(chloromethyl)-4-nitrobenzene
7482
171.58

Chemical Identifiers

100-14-1
171.58
KGCNHWXDPDPSBV-UHFFFAOYSA-N
7482
1-(chloromethyl)-4-nitrobenzene
C7H6ClNO2
MFCD00007374
4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
CHEBI:87406
C1=CC(=CC=C1CCl)[N+](=O)[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H341-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 202-822-7

RTECSNumber : XS9093000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only