missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl chloride, 99%

Catalog No. p-7022823
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1574909 10 g
AAA1574918 50 g
AAA1574930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1574909 Supplier Thermo Scientific Chemicals Supplier No. A1574909
Only null left

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-14-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007374
InChI Key KGCNHWXDPDPSBV-UHFFFAOYSA-N
Synonym 4-nitrobenzyl chloride, 1-chloromethyl-4-nitrobenzene, p-nitrobenzyl chloride, alpha-chloro-4-nitrotoluene, 4-nitrobenzylchloride, benzene, 1-chloromethyl-4-nitro, alpha-chloro-p-nitrotoluene, p-chloromethyl nitrobenzene, 4-chloromethyl nitrobenzene, unii-3bld9lvt3x
PubChem CID 7482
ChEBI CHEBI:87406
IUPAC Name 1-(chloromethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCl)[N+](=O)[O-]

Specifications

Melting Point 70°C to 74°C
Quantity 10 g
UN Number UN3261
Beilstein 387187
Formula Weight 171.58
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl chloride
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H341-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-822-7
RTECSNumber XS9093000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only