accessibility menu, dialog, popup

UserName

ONOCl

CAS RN 100-14-1

IUPAC Name: 1-(chloromethyl)-4-nitrobenzene
Synonyms: 1-(chloromethyl)-4-nitrobenzene
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: KGCNHWXDPDPSBV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CCl)C=C1
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)