missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzyl Acetoacetate 98.0+%, TCI America™

Catalog No. N08755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08755G 5 g
N087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08755G Supplier TCI America Supplier No. N08755G

Chemical Identifiers

CAS 61312-84-3
Molecular Formula C11H11NO5
Molecular Weight (g/mol) 237.211
MDL Number MFCD00799237
InChI Key KQPGVCMZXFBYMM-UHFFFAOYSA-N
Synonym Acetoacetic Acid 4-Nitrobenzyl Ester
PubChem CID 2760918
IUPAC Name (4-nitrophenyl)methyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 44°C
Color Yellow
Quantity 5 g
Formula Weight 237.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl Acetoacetate