missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzonitrile 98.0+%, TCI America™

Catalog No. N017225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N017225G 25 g
N0172250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N017225G Supplier TCI America Supplier No. N017225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 619-72-7
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007279
InChI Key NKJIFDNZPGLLSH-UHFFFAOYSA-N
Synonym benzonitrile, 4-nitro, 4-cyanonitrobenzene, p-nitrobenzonitrile, p-cyanonitrobenzene, benzonitrile, p-nitro, 4-nitrobenzenecarbonitrile, 1-nitro-4-cyanobenzene, 4-nitro-benzonitrile, ccris 2328, 4-nitrobenzonitril
PubChem CID 12090
IUPAC Name 4-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 147°C
Color Yellow
Quantity 25 g
UN Number 3439
Formula Weight 148.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzonitrile
EINECSNumber (3)-1782
RTECSNumber DI4903500
TSCA Yes