missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzoic Acid Potassium Salt 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 205.21 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Nitrobenzoic Acid Potassium Salt |
Safety and Handling
| TSCA | No |