missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzhydrazide, 98+%

Catalog No. AAA1801618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1801618 50 g
AAA1801609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1801618 Supplier Thermo Scientific Chemicals Supplier No. A1801618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 636-97-5 | C7H7N3O3 | 181.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 636-97-5
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007604
InChI Key FKZXYJYTUSGIQE-UHFFFAOYSA-N
Synonym 4-nitrobenzhydrazide, p-nitrobenzhydrazide, 4-nitrobenzoylhydrazine, p-nitrobenzohydrazide, p-nitrobenzoylhydrazide, p-nitrobenzoic hydrazide, p-nitrobenzoic acid hydrazide, p-nitrobenzoyl hydrazine, 4-nitrobenzoic acid hydrazide, 4-nitrobenzoyl hydrazide
PubChem CID 3693744
ChEBI CHEBI:66928
IUPAC Name 4-nitrobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point ∼216°C (decomposition)
Quantity 50 g
Beilstein 519882
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 4-Nitrobenzhydrazide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 211-271-1
RTECSNumber DH5670000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only