missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzaldehyde, 99%

CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol

Supplier:  Thermo Scientific Chemicals 156991000

 View more versions of this product

Catalog No. AC156991000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitrobenzaldehyde
105.0°C to 106.0°C
Authentic
Plastic bottle
O2NC6H4CHO
100 g
14,6587
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and benzene,slightly soluble in ether
[O-][N+](=O)C1=CC=C(C=O)C=C1
151.12
CHEBI:66926
99%
555-16-8
Yellow to Brown
99%
C7H5NO3
MFCD00007346
07,256
p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
BXRFQSNOROATLV-UHFFFAOYSA-N
4-nitrobenzaldehyde
541
151.12
Crystalline Powder

Chemical Identifiers

555-16-8
151.12
BXRFQSNOROATLV-UHFFFAOYSA-N
541
4-nitrobenzaldehyde
C7H5NO3
MFCD00007346
p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
CHEBI:66926
[O-][N+](=O)C1=CC=C(C=O)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 209-084-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only