missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-o-phenylenediamine, 97%

Catalog No. AAL0469336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0469336 500 g
AAL0469322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0469336 Supplier Thermo Scientific Chemicals Supplier No. L0469336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-56-9 | C6H7N3O2 | 153.14 g/mol

4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

Solubility
Slightly soluble in water (1.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 99-56-9
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007724
InChI Key RAUWPNXIALNKQM-UHFFFAOYSA-N
Synonym 4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
PubChem CID 5111791
ChEBI CHEBI:67116
IUPAC Name 4-nitrobenzene-1,2-diamine
SMILES NC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 199°C to 203°C
Quantity 500 g
Beilstein 608106
Merck Index 14,6623
Solubility Information Slightly soluble in water (1.2g/L at 20°C).
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Nitro-o-phenylenediamine
Hazard Category H302-H317-H340
Hazard Statement GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 202-766-3
RTECSNumber ST2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only