missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-m-phenylenediamine, 95%

Catalog No. AAL1216618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1216618 50 g
AAL1216609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1216618 Supplier Thermo Scientific Chemicals Supplier No. L1216618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 5131-58-8 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5131-58-8
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00025289
InChI Key DPIZKMGPXNXSGL-UHFFFAOYSA-N
Synonym 4-nitro-m-phenylenediamine, 4-nitro-1,3-phenylenediamine, 2,4-diaminonitrobenzene, 1,3-benzenediamine, 4-nitro, 4-nitro-1,3-benzenediamine, 1,3-diamino-4-nitrobenzene, m-phenylenediamine, 4-nitro, 3-amino-4-nitrophenylamine, 4-nitro-1,3-fenylendiamin, unii-0d5u5ew62z
PubChem CID 21208
IUPAC Name 4-nitrobenzene-1,3-diamine
SMILES NC1=CC=C(C(N)=C1)[N+]([O-])=O

Specifications

Melting Point 156°C to 160°C
Quantity 50 g
Beilstein 2094904
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 4-Nitro-m-phenylenediamine
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H301-H341-H315-H319
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 225-876-3
RTECSNumber ST2910000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only