missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-L-phenylalanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7134006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08781G 1 g
N08785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08781G Supplier TCI America Supplier No. N08781G

Chemical Identifiers

CAS 17193-40-7
Molecular Formula C10H13ClN2O4
Molecular Weight (g/mol) 260.674
MDL Number MFCD00135754
InChI Key BTHMRXRBXYHLRA-FVGYRXGTSA-N
Synonym 4-nitro-l-phenylalanine methyl ester hydrochloride, 4-nitro-l-phenylalanine methyl ester hcl, h-phe 4-no2-ome.hcl, l-4-nitrophenylalanine methyl ester hydrochloride, s-methyl 2-amino-3-4-nitrophenyl propanoate hydrochloride, s-+-4-nitrophenylalanine methyl ester hydrochloride, s-4-nitrophenylalanine methyl ester hydrochloride, h-phe 4'-no2-ome hcl, methyl 2s-2-amino-3-4-nitrophenyl propanoate hydrochloride, p-nitrophenylalanine methyl ester hydrochloride
PubChem CID 11715866
IUPAC Name methyl (2S)-2-amino-3-(4-nitrophenyl)propanoate;hydrochloride
SMILES COC(=O)C(CC1=CC=C(C=C1)[N+](=O)[O-])N.Cl

Specifications

Melting Point 224°C
Color Yellow
Quantity 1 g
Formula Weight 260.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-L-phenylalanine Methyl Ester Hydrochloride