missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2,3-lutidine N-oxide, 97%, Thermo Scientific™

Catalog No. AC441980050
Change view
Click to view available options
Quantité:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
AC441980050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC441980050 Supplier Thermo Scientific Chemicals Supplier No. 441980050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37699-43-7
Formule moléculaire C7H8N2O3
Poids moléculaire (g/mol) 168.15
Numéro MDL MFCD00065172
Clé InChI CFMTVTYBZMKULI-UHFFFAOYSA-N
Synonyme 2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium
PubChem CID 148223
Nom de l’IUPAC 2,3-dimethyl-4-nitro-1-oxidopyridin-1-ium
SOURIRES CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O

Specifications

Nom ou substance chimique 4-Nitro-2, 3-lutidine N-oxide
CAS % minimum 96.0
CAS % maximal 100.0
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96% min. (HPLC)
Quantité 5g
Poids de formule 168.15
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only