Learn More
Chemglass Inc Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 150mm Schott Flange
Numéro de catalogue. CG194103
Encompass
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 150mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.
Numéro de catalogue. CG194103 Fournisseur Chemglass Inc Code fournisseur CG194103
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Reaction Vessel Lid, 4-Neck, 24/40 Center Neck, 3-24/40 Side Necks, 150mm Schott Flange. Four neck vessel lid to fit CG-1920 through CG-1936 vessels. Lid has a flat ground surface allowing it to be greased or used with an o-ring. Note: Four neck lids are available with joint sizes and configurations other than those listed. Please call for details.