missing translation for 'onlineSavingsMsg'
Learn More

4-n-Pentyloxyphenylacetylene, 98+%

Catalog No. AAH2755509
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2755509 10 g
AAH2755503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2755509 Supplier Thermo Scientific Chemicals Supplier No. H2755509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79887-16-4 | C13H16O | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79887-16-4
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00173877
InChI Key MKSWQHOPSDCVMS-UHFFFAOYSA-N
Synonym 1-eth-1-ynyl-4-pentyloxy benzene, 1-ethynyl-4-pentyloxy benzene, p-ethynyl pentyloxy benzene, 4-n-pentyloxyphenylacetylene, 4-pentoxyphenylacetylene, acmc-1bl8i, 1-4-pentoxyphenyl ethyne, 1-ethynyl-4-pentyloxybenzene, 4-n-pentyloxy-phenylacetylene, 4-n-pentyloxyphenyl acetylene
PubChem CID 2775119
IUPAC Name 1-ethynyl-4-pentoxybenzene
SMILES CCCCCOC1=CC=C(C=C1)C#C

Specifications

Boiling Point 113°C to 115°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.531
Quantity 10 g
Formula Weight 188.27
Percent Purity 99%
Chemical Name or Material 4-n-Pentyloxyphenylacetylene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only