missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylbenzoic Acid 97.0+%, TCI America™

Catalog No. O01375G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01375G 5 g
O01371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O01375G Supplier TCI America Supplier No. O01375G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3575-31-3
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00042649
InChI Key ZQLDNJKHLQOJGE-UHFFFAOYSA-N
Synonym 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984
PubChem CID 19147
IUPAC Name 4-octylbenzoic acid
SMILES CCCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 113°C
Color White
Quantity 5 g
Formula Weight 234.34
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octylbenzoic Acid
EINECSNumber (3)-3544
RTECSNumber DH5945000
TSCA Yes