missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 98.0+%, TCI America™

Catalog No. O0431200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0431200MG 200 mg
O04311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0431200MG Supplier TCI America Supplier No. O0431200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141029-75-6
Molecular Formula C16H21NS2
Molecular Weight (g/mol) 291.47
MDL Number MFCD26793506
InChI Key GGCJLBJHTUIOES-UHFFFAOYSA-N
PubChem CID 11033644
IUPAC Name 7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
SMILES CCCCCCCCN1C2=C(SC=C2)C2=C1C=CS2

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 200 mg
Formula Weight 291.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octyl-4H-dithieno[3,2-b:2′,3′-d]pyrrole
TSCA No
Recommended Storage Refrigerator