missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonyloxyaniline, 98%

CAS: 50262-67-4 | C15H25NO | 235.371 g/mol

$126.58 - $443.10

Chemical Identifiers

CAS 50262-67-4
Molecular Formula C15H25NO
Molecular Weight (g/mol) 235.371
MDL Number MFCD00043622
InChI Key JQLBBFVOAHUASD-UHFFFAOYSA-N
Synonym p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy
PubChem CID 142698
IUPAC Name 4-nonoxyaniline
SMILES CCCCCCCCCOC1=CC=C(C=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0912604
View Documents
Thermo Scientific Chemicals
L0912604
2 g
Each for $126.58
Only null left
 
AAL0912609
View Documents
Thermo Scientific Chemicals
L0912609
10 g
Each for $443.10
Only null left
 
Description

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

50262-67-4
235.371
JQLBBFVOAHUASD-UHFFFAOYSA-N
142698
CCCCCCCCCOC1=CC=C(C=C1)N
C15H25NO
MFCD00043622
p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy
4-nonoxyaniline

Specifications

50262-67-4
176°C to 179°C (2 mmHg)
MFCD00043622
UN2811
p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy
JQLBBFVOAHUASD-UHFFFAOYSA-N
4-nonoxyaniline
142698
98%
36°C to 42°C
C15H25NO
2 g
4232321
Insoluble in water.
CCCCCCCCCOC1=CC=C(C=C1)N
235.371
235.37
4-n-Nonyloxyaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only