Learn More
4-n-Nonyloxyaniline, 98%
CAS: 50262-67-4 | C15H25NO | 235.371 g/mol
Supplier: Thermo Scientific Chemicals L0912609
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 4-n-Nonyloxyaniline | |
| 36°C to 42°C | |
| C15H25NO | |
| 10 g | |
| 4232321 | |
| Insoluble in water. | |
| CCCCCCCCCOC1=CC=C(C=C1)N | |
| 235.371 | |
| 235.37 |
| 50262-67-4 | |
| 176°C to 179°C (2 mmHg) | |
| MFCD00043622 | |
| UN2811 | |
| p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy | |
| JQLBBFVOAHUASD-UHFFFAOYSA-N | |
| 4-nonoxyaniline | |
| 142698 | |
| 98% |
Chemical Identifiers
| 50262-67-4 | |
| 235.371 | |
| JQLBBFVOAHUASD-UHFFFAOYSA-N | |
| 142698 | |
| CCCCCCCCCOC1=CC=C(C=C1)N |
| C15H25NO | |
| MFCD00043622 | |
| p-nonyloxyaniline, 4-n-nonyloxyaniline, 4-nonyloxyaniline, 4-nonyloxy aniline, 4-nonyloxyphenylamine, 4-nonyloxy aniline #, benzenamine,4-nonyloxy | |
| 4-nonoxyaniline |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only