CAS RN 50262-67-4
IUPAC Name:
4-(nonyloxy)aniline
Synonyms:
4-(nonyloxy)aniline
Molecular Weight (g/mol):
235.37
Molecular Formula:
C15H25NO
InChi Key:
JQLBBFVOAHUASD-UHFFFAOYSA-N
SMILES:
CCCCCCCCCOC1=CC=C(N)C=C1
1
–
1
of
1
results
1
Promotions Available
4-n-Nonyloxyaniline, 98%
4-n-Nonyloxyaniline, 98%
CAS: 50262-67-4 Molecular Formula: C15H25NO Molecular Weight (g/mol): 235.371 MDL Number: MFCD00043622 InChI Key: JQLBBFVOAHUASD-UHFFFAOYSA-N Synonym: p-nonyloxyaniline,4-n-nonyloxyaniline,4-nonyloxyaniline,4-nonyloxy aniline,4-nonyloxyphenylamine,4-nonyloxy aniline #,benzenamine,4-nonyloxy PubChem CID: 142698 IUPAC Name: 4-nonoxyaniline SMILES: CCCCCCCCCOC1=CC=C(C=C1)N