missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Dimethyl)aniline magnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC450610500
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450610500 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450610500 Supplier Thermo Scientific Chemicals Supplier No. 450610500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7353-91-5 | C8H10BrMgN | 224.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7353-91-5
Molecular Formula C8H10BrMgN
Molecular Weight (g/mol) 224.38
MDL Number MFCD00672006
InChI Key OWWWKAXERYRWAU-UHFFFAOYSA-M
Synonym grignard reagent
PubChem CID 10933129
IUPAC Name 4-(bromomagnesio)-N,N-dimethylaniline
SMILES CN(C)C1=CC=C([Mg]Br)C=C1

Specifications

Packaging AcroSeal™ Glass bottle
Density 0.9510g/mL
Flash Point −17°C
Specific Gravity 0.951
Linear Formula (CH3)2NC6H4MgBr
Solubility Reacts in water
Quantity 50 mL
Formula Weight 224.38
Chemical Name or Material 4-(N, N-Dimethyl)aniline magnesium bromide

RUO – Research Use Only