missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxybenzoyl chloride, 98%

Catalog No. AAA1336706
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1336706 5 g
AAA1336722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1336706 Supplier Thermo Scientific Chemicals Supplier No. A1336706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 39649-71-3 | C13H17ClO2 | 240.727 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39649-71-3
Molecular Formula C13H17ClO2
Molecular Weight (g/mol) 240.727
MDL Number MFCD00000690
InChI Key DQQOONVCLQZWOY-UHFFFAOYSA-N
PubChem CID 170235
IUPAC Name 4-hexoxybenzoyl chloride
SMILES CCCCCCOC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.082
Boiling Point 203°C to 205°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5383
Quantity 5 g
UN Number UN3265
Beilstein 2210619
Sensitivity Moisture sensitive
Synonym 4-n-hexyloxybenzoyl chloride, 4-hexyloxybenzoyl chloride, 4-hexyloxy benzoyl chloride, benzoyl chloride, 4-hexyloxy, p-hexyloxybenzoyl chloride, 4-hexyloxybenzoylchloride, 4-hexyloxy-benzoyl chloride, 4-hexyloxybenzoic acid chloride, 4-n-hexyl-oxybenzoyl chloride, benzoyl chloride,4-hexyloxy
Formula Weight 240.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexyloxybenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-561-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only