missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzoyl chloride, 98%

Numéro de catalogue. p-7022755
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1565914 25 g
AAA1565922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1565914 Fournisseur Thermo Scientific Chemicals Code fournisseur A1565914
Il en reste null

CAS: 50606-95-6 | C13H17ClO | 224.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 50606-95-6
Molecular Formula C13H17ClO
Molecular Weight (g/mol) 224.73
MDL Number MFCD00000701
InChI Key XRAHLPNMIIAEPP-UHFFFAOYSA-N
Synonym 4-n-hexylbenzoyl chloride, benzoyl chloride, 4-hexyl, p-hexylbenzoyl chloride, 4-n-hexylbenzoylchloride, 4-n-hexyl-benzoyl chloride, 4-hexylbenzoyl chloride, para-n-hexyl benzoic acid chloride, 4-hexylbenzene-1-carbonyl chloride
PubChem CID 170863
IUPAC Name 4-hexylbenzoyl chloride
SMILES CCCCCCC1=CC=C(C=C1)C(Cl)=O

Spécifications

Density 1.029
Boiling Point 176°C to 177°C (22 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5256
Quantity 25 g
UN Number UN3265
Beilstein 2208938
Sensitivity Moisture sensitive
Formula Weight 224.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexylbenzoyl chloride
Afficher plus de résultats Afficher moins de résultats
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 256-647-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only