missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylbenzoyl chloride, 98%

Catalog No. AAA1723414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1723414 25 g
AAA1723406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1723414 Supplier Thermo Scientific Chemicals Supplier No. A1723414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54256-43-8 | C17H25ClO | 280.836 g/mol

Employed as a pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical and chemical intermediate.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.

Chemical Identifiers

CAS 54256-43-8
Molecular Formula C17H25ClO
Molecular Weight (g/mol) 280.836
MDL Number MFCD00041724
InChI Key HEWLDHSFOQXCSI-UHFFFAOYSA-N
PubChem CID 104714
IUPAC Name 4-decylbenzoyl chloride
SMILES CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Boiling Point 171°C to 173°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride
Solubility Information Reacts with water.
Formula Weight 280.84
Percent Purity 98%
Chemical Name or Material 4-n-Decylbenzoyl chloride
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 259-046-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only