missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzonitrile, 97%, Thermo Scientific™

Catalog No. AAA1657014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1657014 25 g
AAA1657006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1657014 Supplier Thermo Scientific Chemicals Supplier No. A1657014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 5203-14-5
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00043482
InChI Key RRGQINKVTNAIBB-UHFFFAOYSA-N
Synonym p-butoxybenzonitrile, 4-n-butoxybenzonitrile, benzonitrile, 4-butoxy, 4-n-butoxy benzonitrile, 4-butoxy-benzonitrile, 4-butoxybenzenecarbonitrile, pubchem7926, acmc-209kxf, 4-butoxybenzonitrile
PubChem CID 138435
IUPAC Name 4-butoxybenzonitrile
SMILES CCCCOC1=CC=C(C=C1)C#N

Specifications

Density 0.999
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.5255
Quantity 25 g
Beilstein 2834664
Formula Weight 175.23
Percent Purity 97%
Chemical Name or Material 4-n-Butoxybenzonitrile
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures