missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butoxybenzeneboronic acid, 98%

Catalog No. AAH5286506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5286506 5 g
AAH5286503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5286506 Supplier Thermo Scientific Chemicals Supplier No. H5286506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105365-51-3 | C10H15BO3 | 194.04 g/mol

Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 105365-51-3
Molecular Formula C10H15BO3
Molecular Weight (g/mol) 194.04
MDL Number MFCD03427054
InChI Key QUPFQMXWFNJUNJ-UHFFFAOYSA-N
Synonym 4-n-butoxyphenylboronic acid, 4-butoxyphenyl boronic acid, 4-n-butoxyphenyl boronic acid, p-butoxyphenylboronic acid, 4-butyloxyphenylboronic acid, 4-n-butoxy benzeneboronic acid, 4-butoxymethylboronic acid, boronic acid, 4-butoxyphenyl, pubchem9549, 4-butoxybenzeneboronic acid
PubChem CID 3836310
IUPAC Name (4-butoxyphenyl)boronic acid
SMILES CCCCOC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 102°C to 104°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 4-n-Butoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only