missing translation for 'onlineSavingsMsg'
Learn More

4-[(N-BOC)aminomethyl]aniline, 97%

Catalog No. AC426200010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC426200010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC426200010 Supplier Thermo Scientific Chemicals Supplier No. 426200010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94838-55-8 | C12H18N2O2 | 222.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94838-55-8
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD03001716
InChI Key UXWQXBSQQHAGMG-UHFFFAOYSA-N
Synonym 4-n-boc-aminomethyl aniline, tert-butyl 4-aminobenzylcarbamate, tert-butyl n-4-aminophenyl methyl carbamate, tert-butyl 4-aminobenzyl carbamate, 4-n-boc aminomethyl aniline, 4-aminobenzyl carbamic acid tert-butyl ester, carbamic acid, 4-aminophenyl methyl-, 1,1-dimethylethyl ester, n-4-aminophenyl methyl tert-butoxy carboxamide, butylaminobenzylcarbamate, p-bocnhch2c6h4nh2
PubChem CID 2756041
IUPAC Name tert-butyl N-[(4-aminophenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(N)C=C1

Specifications

Melting Point 75°C to 80°C
CAS Min % 96.0
CAS Max % 100.0
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula NH2C6H4CH2NHCO2C(CH3)3
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-[(N-BOC)aminomethyl]aniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only