missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Morpholinylcarbonyl Chloride 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 138°C |
| Quantity | 25 g |
| UN Number | 3265 |
| Formula Weight | 149.57 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Morpholinylcarbonyl Chloride |
Safety and Handling
| EINECSNumber | (5)-3787 |
| TSCA | Yes |