missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric acid, 99%

Catalog No. AAA1540522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1540522 100 g
AAA1540506 5 g
AAA1540514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1540522 Supplier Thermo Scientific Chemicals Supplier No. A1540522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 646-07-1 | C6H12O2 | 116.16 g/mol

4-Methylvaleric acid has been used in asymmetric synthesis of (4S)-4-isopropyl-3-[(2?S)-2?,4?-dimethyl-valeryl)]-2-oxazolidinone. It is a versatile building block used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors and antagonists. It is involved in the synthesis of novel series of high affinity, functionally potent antagonists of the CCR1 receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylvaleric acid has been used in asymmetric synthesis of (4S)-4-isopropyl-3-[(2′S)-2′,4′-dimethyl-valeryl)]-2-oxazolidinone. It is a versatile building block used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors and antagonists. It is involved in the synthesis of novel series of high affinity, functionally potent antagonists of the CCR1 receptor.

Solubility
Soluble in alcohol. Slightly soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents, bases.

Chemical Identifiers

CAS 646-07-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002803
InChI Key FGKJLKRYENPLQH-UHFFFAOYSA-N
Synonym 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid
PubChem CID 12587
ChEBI CHEBI:74903
IUPAC Name 4-methylpentanoic acid
SMILES CC(C)CCC(=O)O

Specifications

Melting Point -35°C
Density 0.923
Boiling Point 199°C to 201°C
Flash Point 97°C (206°F)
Odor Unpleasant
Refractive Index 1.4146
Quantity 100 g
UN Number UN2922
Beilstein 1741912
Solubility Information Soluble in alcohol. Slightly soluble in water.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 4-Methylvaleric acid
Show More Show Less
Hazard Category H311-H314
Hazard Statement GHS H Statement
H311-H314-H318
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 211-464-0
RTECSNumber NR2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only