missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-glucuronide dihydrate, 98%

Catalog No. AC337220250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC337220250 25 g
AC337220010 1 g
AC337220050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC337220250 Supplier Thermo Scientific Chemicals Supplier No. 337220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6160-80-1 | C16H16O9·2H2O | 388.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6160-80-1
Molecular Formula C16H16O9·2H2O
Molecular Weight (g/mol) 388.33
InChI Key ARQXEQLMMNGFDU-JHZZJYKESA-N
Synonym 4-methylumbelliferyl-beta-d-glucuronide, 4-methylumbelliferyl glucuronide, 4-methylumbelliferone glucuronide, mug trihydrate, 4-methylumbelliferyl beta-glucuronide, 4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
PubChem CID 91553
ChEBI CHEBI:1904
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O

Specifications

Melting Point 102.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Specific Rotation Condition −105.00 (30.00°C c=0.25, water)
Specific Rotation -105
Solubility Information (c=1,H2O at 60°C) Clear colorless solution
Formula Weight 388.33
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-β-D-glucuronide dihydrate
EINECSNumber 228-186-

RUO – Research Use Only