missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-beta-D-xylopyranoside, 99%

Catalog No. AC337272500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC337272500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC337272500 Supplier Thermo Scientific Chemicals Supplier No. 337272500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6734-33-4 | C15H16O7 | 308.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6734-33-4
Molecular Formula C15H16O7
Molecular Weight (g/mol) 308.29
MDL Number MFCD00037607
InChI Key JWIYLOHVJDJZOQ-KAOXEZKKSA-N
Synonym 4-methylumbelliferyl b-d-xylopyranoside, 4-methylumbelliferyl beta-d-xyloside, 4-methylumbelliferyl beta-d-xylopyranoside, methylumbelliferyl-beta-d-xyloside, 4-methylumbelliferyl-beta-d-xyloside, 4-methylumbelliferyl a-d-xylopyranoside, 4-methylumbelliferyl-beta-d-xylopyranoside, 2h-1-benzopyran-2-one,4-methyl-7-b-d-xylopyranosyloxy, 2h-1-benzopyran-2-one, 4-methyl-7-beta-d-xylopyranosyloxy, 4-methyl-7-2s,3r,4s,5r-3,4,5-trihydroxyoxan-2-yl oxy chromen-2-one
PubChem CID 92229
IUPAC Name 4-methyl-7-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(CO3)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 223°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition −30° (20°C c=0.3,pyridine)
Specific Rotation -30
Solubility Information Solubility in water: 1g/L. Other solubilities: 50g/L pyridine
Formula Weight 308.29
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl-β-D-xylopyranoside
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 229-784-4

RUO – Research Use Only