missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl beta-D-Cellobioside 98.0+%, TCI America™

Catalog No. M3028500MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3028500MG 500 mg
M3028100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3028500MG Supplier TCI America Supplier No. M3028500MG

Chemical Identifiers

CAS 72626-61-0
Molecular Formula C22H28O13
Molecular Weight (g/mol) 500.453
MDL Number MFCD00063279
InChI Key PRTGXBPFDYMIJH-MKQZUAMYSA-N
PubChem CID 126287
IUPAC Name 7-[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)OC4C(C(C(C(O4)CO)O)O)O)O)O

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 500 mg
Formula Weight 500.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl beta-D-Cellobioside
TSCA No
Recommended Storage Refrigerator