missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Méthylumbelliféryl bêta-D-Cellobioside 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 248°C |
| Color | Blanc-jaune |
| Quantity | 100 mg |
| Formula Weight | 500.45 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 4-Methylumbelliferyl beta-D-Cellobioside |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Réfrigérateur |