missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M3030250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3030250MG 250 mg
M30301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3030250MG Supplier TCI America Supplier No. M3030250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37067-30-4
Molecular Formula C18H21NO8
Molecular Weight (g/mol) 379.37
MDL Number MFCD00065481
InChI Key QCTHLCFVVACBSA-XWEABGILSA-N
Synonym 4-mu-glcnac, 4-methylumbelliferyl-n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-3-yl acetamide, 4-methylumbelliferyl-2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-methylumbelliferyl n-acetyl-, a-d-glucosaminide, 4-methylumbelliferyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxochromen-7-yl oxy oxan-3-yl acetamide, glcnac-b-4mu, 4-methylumbelliferyl 2-acetamido-2-deoxy-, a-d-glucopyranoside, 4-methylumbelliferyl 2-acetamido-2-deoxy-b-d-glucopyranoside
PubChem CID 2733787
IUPAC Name N-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)OC1OC1=CC=C2C(C)=CC(=O)OC2=C1

Specifications

Color White
Quantity 250 mg
Formula Weight 379.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl 2-Acetamido-2-deoxy-beta-D-glucopyranoside
TSCA Yes
Recommended Storage Freezer (-20 °C)