missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-m-cresol, 97%

Catalog No. AAL0735314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0735314 25 g
AAL0735306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0735314 Supplier Thermo Scientific Chemicals Supplier No. L0735314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3120-74-9 | C8H10OS | 154.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3120-74-9
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00045773
InChI Key VKALYYFVKBXHTF-UHFFFAOYSA-N
Synonym 3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
PubChem CID 18391
ChEBI CHEBI:38681
IUPAC Name 3-methyl-4-methylsulfanylphenol
SMILES CC1=C(C=CC(=C1)O)SC

Specifications

Melting Point 54°C to 58°C
Quantity 25 g
Beilstein 1210322
Formula Weight 154.23
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)-m-cresol
Hazard Category H311-H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P312-P332+P313-P361-P363-P501c
EINECSNumber 221-496-7
RTECSNumber GP2225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only