missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzoic Acid 97.0+%, TCI America™

Catalog No. M167525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M167525G Supplier TCI America Supplier No. M167525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13205-48-6
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00002555
InChI Key KWHCPERWLHBLOT-UHFFFAOYSA-N
Synonym 4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa
PubChem CID 83230
IUPAC Name 4-(methylsulfanyl)benzoic acid
SMILES CSC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.21
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)benzoic Acid