Learn More
4-(Methylthio)aniline, 98%
CAS: 104-96-1 | C7H9NS | 139.22 g/mol
Supplier: Thermo Scientific Chemicals L0495014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-(Methylthio)aniline | |
| 1.12 | |
| >110°C (230°F) | |
| C7H9NS | |
| MFCD00007889 | |
| UN2810 | |
| Light sensitive | |
| YKFROQCFVXOUPW-UHFFFAOYSA-N | |
| 4-methylsulfanylaniline | |
| 66038 | |
| 98% |
| 104-96-1 | |
| 272°C to 273°C | |
| Unpleasant | |
| 1.639 | |
| 25 g | |
| 774506 | |
| 4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine | |
| CSC1=CC=C(N)C=C1 | |
| 139.22 | |
| 139.23 |
Chemical Identifiers
| 104-96-1 | |
| 139.22 | |
| YKFROQCFVXOUPW-UHFFFAOYSA-N | |
| 66038 | |
| CSC1=CC=C(N)C=C1 |
| C7H9NS | |
| MFCD00007889 | |
| 4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine | |
| 4-methylsulfanylaniline |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 203-256-3
RTECSNumber : BY6302000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only